missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517711
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with HPK or other members of the GCK family.
GCK Polyclonal antibody specifically detects GCK in Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.Spezifikation
GCK | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P17712, P35557, P52792 | |
GCK | |
Synthetic peptide corresponding to residues surrounding Ser380 of human GCK | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GCK | |
ATP:D-hexose 6-phosphotransferase; FGQTL3; GC kinase; Gck; Gk; GLK; Gls006; glucokinase; glucokinase (hexokinase 4); glucokinase variant 3; GLUKA; hexokinase 4; hexokinase D, pancreatic isozyme; hexokinase type IV; Hexokinase-4; hexokinase-D; HHF3; HK IV; HK4; HKIV; Hlb62; HXKP; LGLK; MEKKK 2; MODY2; RNGK2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
103988, 24385, 2645 | |
-20°C | |
Liquid |