Learn More
GDNF Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Marke: Invitrogen PA19524
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-9524 detects GDNF in human, mouse and rat samples. PA1-9524 has been successfully used in Western blot, ELISA, and Immunohistochemistry procedures. By Western blot, PA1-9524 detects a 24 kDa protein representing GDNF. PA1-9524 immunogen is a synthetic peptide based on the human GDNF protein. Store product at 4oC for up to one week, and -20oC for long term storage.
GDNF Polyclonal antibody specifically detects GDNF in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
GDNF | |
Polyclonal | |
Unconjugated | |
GDNF | |
AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
Chicken | |
IgY fraction | |
RUO | |
14573, 25453, 2668 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P39905, P48540, Q07731 | |
GDNF | |
Synthetic peptide based on the human GDNF protein. | |
500 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |