Learn More
Gemin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544612
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QALAEKVKEA ERDVSLTSLA KLPSETIFVG CEFLHHLLRE WGEELQAVLR Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
Gemin 4 Polyclonal antibody specifically detects Gemin 4 in Human samples. It is validated for Western Blot.Spezifikation
Gemin 4 | |
Polyclonal | |
Unconjugated | |
GEMIN4 | |
4932415L08Rik; AU015359; component of gems 4; DKFZp434B131; DKFZp434D174; gem (nuclear organelle) associated protein 4; gem nuclear organelle associated protein 4; gem-associated protein 4; GEMIN4; Gemin-4; HC56; HCAP1; HCC-associated protein 1; HHRF-1; p97; RGD1563715 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
50628 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P57678 | |
GEMIN4 | |
Synthetic peptide directed towards the N-terminal of human GEMIN4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |