missing translation for 'onlineSavingsMsg'
Learn More

GFAP Mouse, Unlabeled, Clone: 52, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610566

Weitere Details : Netto-Gewicht : 0.01100kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15895519

  • 484.50 € / 1 Set
Verfügbar ab: 20-12-2022
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The cytoskeleton is an array of highly ordered filaments that maintains the shape, structure, and functionality of cells. There are three major types of filaments: actin filaments, microtubules, and intermediate filaments. The distinctions among them are based on protein composition and arrangements. Glial fibrillary acidic

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Spezifikation

Spezifikation

GFAP
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Rat, Mouse, Human, Chicken
IgG1
Western Blot
52
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.