missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 40.1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541320
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: HTKIPDEDES EVTSSASEKL GRQPRGRPHR EAAQDPRGSG SEEQPSAAPS Sequence homology: Human: 100%
Connexin 40.1 Polyclonal antibody specifically detects Connexin 40.1 in Human samples. It is validated for Western Blot.Spezifikation
Connexin 40.1 | |
Polyclonal | |
Unconjugated | |
GJD4 | |
connexin 40.1; connexin40.1; connexin-40.1; CX40.1; Gap junction delta-4 protein; gap junction protein delta 4; gap junction protein, delta 4, 40.1kDa; GJD4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
219770 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96KN9 | |
GJD4 | |
Synthetic peptide directed towards the middle region of human GJD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |