Learn More
anti-HBZ Matched Antibody Pair, Abnova
Marke: Abnova H00003050-AP11.1Set
Weitere Details : Netto-Gewicht : 0.00010kg
Beinhaltet: Rabbit MaxPab7™ affinity purified polyclonal anti-HBZ (100μg); Mouse monoclonal anti-HBZ, IgG1 κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
ζ-globin is an α-like hemoglobin. The ζ-globin polypeptide is synthesized in the yolk sac of the early embryo, while α-globin is produced throughout fetal and adult like. The ζ-globin gene is a member of the human α-globin gene cluster that includes five functional genes and two pseudogenes. The order of genes is: 5′ - ζ - pseudoζ - μ - pseudoα-1 - α-2 -α-1 - θ1 - 3′.
ELISA
Spezifikation
HBZ | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HBZ. | |
HBZ | |
Affinity Purified | |
RUO | |
3050 | |
Human | |
IgG1 |
ELISA | |
Unconjugated | |
HBZ | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-HBZ (100 μg) 2. Detection antibody: mouse monoclonal anti-HBZ, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |