Learn More
HGF Recombinant Polyclonal Antibody (7HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710307
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse, rat, non-human primate and rabbit based on sequence homology. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
HGF Recombinant Polyclonal antibody specifically detects HGF in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
HGF | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF | |
Recombinant protein corresponding to amino acids 27–169 of human HGF | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
7HCLC | |
Unconjugated | |
P14210 | |
Rabbit | |
Protein A | |
RUO | |
3082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |