missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF1A Monoclonal Antibody (H1alpha67), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA116511
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
In Western blot, multiple bands may be seen at 100-120 kDa representing post-translational modification of HIF-1 alpha. For WB, testing on nuclear extracts is recommended.
HIF1A Monoclonal antibody specifically detects HIF1A in Avian, Bovine, Canine, Ferret, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit, Rat samples. It is validated for ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
HIF1A | |
Monoclonal | |
1.0 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide | |
O35800, Q16665, Q61221, Q9XTA5 | |
HIF1A | |
Fusion protein containing residues 432-528 of human HIF-1alpha | |
100 μL | |
Primary | |
Avian, Rat, Bovine, Ferret, Human, Mouse, Non-human Primate, Non-human Primate, Porcine, Rabbit, Rabbit | |
Antibody | |
IgG2b |
ChIP Assay, ELISA, Flow Cytometry, Gel Shift, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
H1alpha67 | |
Unconjugated | |
HIF1A | |
AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L home | |
Mouse | |
Protein A | |
RUO | |
100009579, 15251, 281814, 29560, 3091, 396696, 443519, 480348 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |