missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546708
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: SQQNLFDNKF DDIFGSSFSS DPFNFNSQNG VNKDEKDHLI ERLYREISGL
HIP1 Polyclonal antibody specifically detects HIP1 in Human samples. It is validated for Western Blot.Spezifikation
HIP1 | |
Polyclonal | |
Unconjugated | |
HIP1 | |
2610109B09Rik; A930014B11Rik; E130315I21Rik; HIP I; hip1; HIP-1; HIP-I; huntingtin interacting protein 1; huntingtin-interacting protein 1; Huntingtin-interacting protein I; ILWEQ; Kiaa4113; mKIAA4113; SHON; SHONbeta; SHONgamma | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3092 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00291 | |
HIP1 | |
Synthetic peptide directed towards the N-terminal of human HIP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |