Learn More
HIP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530058
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HepG2, HCT116, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (98%), Pig (100%), Chicken (99%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HIP2 Polyclonal antibody specifically detects HIP2 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
HIP2 | |
Polyclonal | |
Unconjugated | |
UBE2K | |
AW492011; D5Ertd601e; DKFZp564C1216; DKFZp686J24237; E2 ubiquitin-conjugating enzyme K; E2(25K); E2-25k; HIP2; HIP-2; huntingtin interacting protein 2; huntingtin-interacting protein 2; HYPG; LIG; UBC1; UBE2K; Ubiquitin carrier protein; ubiquitin conjugating enzyme E2 K; ubiquitin conjugating enzyme E2K; ubiquitin-conjugating enzyme E2 K; Ubiquitin-conjugating enzyme E2(25K); ubiquitin-conjugating enzyme E2-25 KDA; ubiquitin-conjugating enzyme E2-25K; ubiquitin-conjugating enzyme E2K; ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast); ubiquitin-protein ligase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3093, 53323 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P61086, P61087 | |
UBE2K | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 167 of Human HIP2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |