Learn More
PPIP5K2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529340
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Raji, K562. Predicted reactivity: Mouse (99%), Rat (97%), Xenopus laevis (94%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PPIP5K2 Polyclonal antibody specifically detects PPIP5K2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PPIP5K2 | |
Polyclonal | |
Unconjugated | |
PPIP5K2 | |
AW555814; CFAP160; D330021B20; diphosphoinositol pentakisphosphate kinase 2; Hisppd1; histidine acid phosphatase domain containing 1; histidine acid phosphatase domain-containing protein 1; hsVIP2; inositol heptaphosphate kinase 2; inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; InsP6 and PP-IP5 kinase 2; IP7K2; KIAA0433; LOW QUALITY PROTEIN: inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 2; mKIAA0433; mmVIP2; PPIP5K2; VIP1 homolog 2; Vip2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23262 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43314 | |
PPIP5K2 | |
Recombinant fragment corresponding to a region within amino acids 5 and 341 of Human HISPPD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |