missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1.2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532009
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HeLa nucleus. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Histone H1.2 Polyclonal antibody specifically detects Histone H1.2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Histone H1.2 | |
Polyclonal | |
Unconjugated | |
H1-2 | |
0610008C09Rik; CTL-1; H1 histone family, member 2; H1 VAR.1; H1.2; H1.2 linker histone, cluster member; H1-2; H1C; H1f2; H1s-1; H1var1; His1a; Hist1h1c; histone 1, H1c; histone cluster 1, H1c; histone H1; histone H1.1; histone H1.2; Histone H1c; histone H1d; Histone H1s-1; LOC684681; MGC3992; similar to Histone H1.2 (H1 VAR.1) (H1c) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3006 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.88 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P16403 | |
H1-2, HIST1H1C | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 184 of Human Histone H1.2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |