Learn More
H3K9acK14ac Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532028
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3, JC, BCL-1, HeLa (0.4 μM Trichostatin treatment for 18 hr). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
H3K9acK14ac Polyclonal antibody specifically detects H3K9acK14ac in Human, Mouse samples. It is validated for Dot Blot, Immunocytochemistry, Western Blot.Spezifikation
H3K9acK14ac | |
Polyclonal | |
Unconjugated | |
H3C15 | |
Acetyl H3; Acetyl Histone H3; Acetyl-H3 K9 K14; Acetyl-H3 Lys9 Lys14; Acetyl-Histone H3 K9 K14; Acetyl-Histone H3 Lys9 Lys14; B0035.7; CELE_T10C6.12; CG31613; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; Dmel\CG31613; Dmel_CG31613; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; h3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3 K9acK14ac; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126961, 260423, 319150, 319152, 333932, 360198, 8350, 8356, 8357, 8358, 8968, 97114 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Dot Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P68431, P68433, P84228, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7 | |
Synthetic Acetyl-peptide corresponding to amino acids residues surrounding the Acetyl Lys9, Lys14 of Histone H3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |