Learn More
HS3ST1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540647
Weitere Details : Netto-Gewicht : 0.00010kg
Description
Peptide sequence: TKGFYCLRDS GRDRCLHESK GRAHPQVDPK LLNKLHEYFH EPNKKFFELV Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%
HS3ST1 Polyclonal antibody specifically detects HS3ST1 in Human samples. It is validated for Western Blot.Specifications
HS3ST1 | |
Polyclonal | |
Unconjugated | |
HS3ST1 | |
3OST; 3-Ost; 3OST1; 3-OST-1; D5Wsu110e; h3-OST-1; heparan sulfate; heparan sulfate (glucosamine) 3-O-sulfotransferase 1; heparan sulfate 3-O-sulfotransferase 1; heparan sulfate 3-O-sulfotransferase-1; heparan sulfate 3-O-sulfotransferase-1 precursor; heparan sulfate D-glucosaminyl 3-O-sulfotransferase 1; Heparan sulfate glucosamine 3-O-sulfotransferase 1; heparan sulfate-glucosamine 3-sulfotransferase 1; heparin-glucosamine 3-O-sulfotransferase; HS3ST1; Hsg3ost | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9957 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14792 | |
HS3ST1 | |
Synthetic peptide directed towards the middle region of human HS3ST1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |