missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSH2D Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519280
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
T-cell activation requires 2 signals: recognition of antigen by the T-cell receptor and a costimulatory signal provided primarily by CD28 in naive T cells. HSH2 is a target of both of these signaling pathways.Spezifikation
HSH2D | |
Polyclonal | |
Unconjugated | |
HSH2D | |
Adaptor in lymphocytes of unknown function X; ALX; FLJ14886; hematopoietic SH2 domain containing; hematopoietic SH2 domain-containing protein; Hematopoietic SH2 protein; HSH2; HSH2D | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
84941 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96JZ2 | |
HSH2D | |
Synthetic peptide sequence (HSHVGYTLSYKAQ) corresponding to the internal amino acids of HSH2D | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |