Learn More
HSP90 beta Monoclonal Antibody (MBH90B), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110373
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody recognizes the EEVHHG epitope within the N-terminal part of Hsp90 beta, a ubiquitously expressed protein with calculated MW of 83.3 kDa which migrates, however, as a 90 kDa band under reducing SDS-PAGE conditions.
HSP90 beta Monoclonal antibody specifically detects HSP90 beta in Bovine, Human, Mouse samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
HSP90 beta | |
Monoclonal | |
1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P08238, P11499, Q76LV1 | |
Hsp90ab1 | |
Peptide corresponding to the EEVHHGEEEVEC sequence within N-terminal part of human Hsp90. | |
100 μg | |
Primary | |
Bovine, Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
MBH90B | |
Unconjugated | |
Hsp90ab1 | |
90kDa; 90-kDa heat shock protein beta; AL022974; C81438; D6S182; heat shock 84 kDa; heat shock 90 kDa protein; heat shock 90kD protein 1, beta; heat shock 90kDa protein 1, beta; heat shock cognate protein HSP 90-beta; Heat shock protein; heat shock protein 1, beta; heat shock protein 90 alpha (cytosolic), class B member 1; heat shock protein 90 alpha family class B member 1; heat shock protein 90 beta; heat shock protein 90 kDa; heat shock protein 90 kDa alpha class B member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-beta; heat shock protein, 84 kDa 1; HSP; HSP 84; HSP 90; Hsp84; Hsp84-1; Hsp90; hsp90 beta; HSP90AB1; HSP90B; HSP90-beta; HSPC2; HSPCB; retinal degeneration slow protein; RP1-302G2.1; TSTA; Tumor-specific transplantation 84 kDa antigen; Unknown (protein for IMAGE:7943063) | |
Mouse | |
Protein A | |
RUO | |
15516, 3326, 767874 | |
4° C, do not freeze | |
Liquid |