missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELAVL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA536157
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.
ELAVL2 Polyclonal antibody specifically detects ELAVL2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ELAVL2 | |
Polyclonal | |
Unconjugated | |
ELAVL2 | |
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B); ELAV like 4; ELAV like neuron-specific RNA binding protein 2; Elav2; ELAVL 2; ELAVL 4; ELAVL2; ELAV-like neuronal protein 1; ELAV-like neuronal protein 1 isoform Hel-N2; ELAV-like protein 2; embryonic lethal, abnormal vision-like 2; HELN1; HEL-N1; Hu antigen B; Hu antigen D; hu-antigen B; HUB; HUD; mel-N1; Nervous system-specific RNA-binding protein Hel-N1; nervous system-specific RNA-binding protein Mel-N1; RNA binding protein HuB; zgc:91918 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15569, 1993, 286973 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12926, Q60899, Q8CH84 | |
ELAVL2 | |
Synthetic peptide corresponding to amino acids 50-100 of human Hel-N1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |