missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF-6 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
N.V.
Spezifikation
Antigen | FGF-6 |
---|---|
Konzentration | LYOPH |
Anwendungen | Western Blot, Neutralization |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Beschreibung
FGF-6 Polyclonal specifically detects FGF-6 in Human samples. It is validated for Western Blot, Neutralization.Spezifikation
FGF-6 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2 | |
FGF6 | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Polyclonal | |
Goat | |
Human | |
2251 | |
IgG | |
Affinity Purified |