missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Follicle Stimulating Hormone Mouse anti-Bovine, Human, Clone: FSH03, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-12144 targets Human Follicle Stimulating Hormone in IHC (P) applications and shows reactivity with Bovine and Human samples. The MA5-12144 immunogen is purified human FSH-beta.
Follicle-stimulatine hormone (FSH) is a member of the glycoprotein hormone family. Members of this family consist of an identical alpha subunit and a hormone-specific beta subunit. Alternative splicing results in two transcript variants encoding the same protein. FSH is secreted by gonadotropic cells in the pituitary gland and in conjunction with luteinizing hormone, induces egg and sperm production.Spezifikation
Follicle Stimulating Hormone | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P04837, P01225 | |
FSHB | |
Purified human FSH-beta | |
500 μL | |
Primary | |
Bovine, Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
FSH03 | |
Unconjugated | |
FSHB | |
FSH-B, FSH-beta, follicle-stimulating hormone beta subunit, follitropin beta chain, follitropin subunit beta, follitropin, beta chain | |
Mouse | |
Protein G | |
RUO | |
2488, 281171 | |
4° C | |
Liquid |