missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Humanin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141610
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Suggested positive control: recombinant fusion protein.
Humanin Polyclonal antibody specifically detects Humanin in Human samples. It is validated for ELISA, Western Blot.Spezifikation
Humanin | |
Polyclonal | |
Unconjugated | |
MT-RNR2 | |
BCL2L4; HN1; Humanin-like 1; humanin-like protein 1; mitochondrially encoded 16S RNA; MTRNR2; MT-RNR2; MTRNR2L1; MT-RNR2-like 1; MTRNR2-like 1; MT-RNR2-like protein 1; RNR2 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
Conc. Not Determined | |
whole serum with 50% glycerol and 0.025% sodium azide | |
Q8IVG9 | |
MT-RNR2 | |
Synthetic peptide corresponding to residues 12-24 of human humanin. | |
RUO | |
4550 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |