missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HuR Monoclonal Antibody (19F12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A21277
Additional Details : Netto-Gewicht : 0.00525kg
Description
HuR Monoclonal antibody specifically detects HuR in Human samples. It is validated for Western Blot.Specifications
HuR | |
Monoclonal | |
Unconjugated | |
ELAVL1 | |
2410055N02Rik; ELAV (embryonic lethal, abnormal vision)-like 1 (Hu antigen R); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV like 1; ELAV like RNA binding protein 1; ELAV like RNA binding protein 1a; ELAV1; Elavl1; elavl1 protein; elavl1a; elav-like generic protein; ELAV-like protein 1; ELAV-like protein 1; ELAV-like protein 1a; elav-related A; Elra; embryonic lethal, abnormal vision, drosophila, homolog-like 1; embryonic lethal, abnormal vision-related A; etID309757.2; fa25f08; fb59a02; fc60g02; Hu antigen R; Hua; HU-antigen A; hu-antigen R; Human antigen R; HuR; melG; ribonucleoprotein; W91709; wu:fa25f08; wu:fb59a02; wu:fc60g02; zHuA | |
Mouse | |
purified | |
RUO | |
1994 | |
-20°C | |
Liquid |
Western Blot | |
19F12 | |
PBS with 0.1% sodium azide; pH 7.4 | |
Q15717 | |
ELAVL1 | |
Human HuR mRNA-binding protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |