missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA536085
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 309 kDa. Purity is >95% by SDS-PAGE.
HYD Polyclonal antibody specifically detects HYD in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HYD | |
Polyclonal | |
Unconjugated | |
UBR5 | |
CH D1S3362; DD5; E3 identified by differential display; E3 ubiquitin-protein ligase UBR5; E3 ubiquitin-protein ligase, HECT domain-containing 1; ECGF1; Edd; Edd1; EDG1; EDG-1; extraembryonic development; HECT-type E3 ubiquitin transferase UBR5; hHYD; HYD; Hyperplastic discs protein homolog; KIAA0896; Progestin-induced protein; Rat100; S1P receptor 1; S1P receptor Edg-1; S1P1; S1PR1; ubiquitin protein ligase E3 component n-recognin 5; Ubr5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117060, 51366, 70790 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O95071, Q62671, Q80TP3 | |
UBR5 | |
Synthetic peptide corresponding to amino acids 1-50 of human HYD | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |