Learn More
IL-10 Rat anti-Human, Unlabeled, Clone: JES3-19F1, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 554704
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The JES3-19F1 antibody reacts with human (IL-10). The immunogen used to generate the JES3-19F1 hybridoma was recombinant human IL-10 expressed in COS cells. This is a neutralizing antibody. This antibody also reacts with viral IL-10.
Host Species: Rat
Clone: JES3-19F1
Isotype: IgG2a κ
Species Reactivity: Human
Immunogen: Recombinant Human IL-10
Functional Assay, Neutralization
Spezifikation
IL-10 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Human IL-10 | |
0.1 mg | |
Immunology | |
Human, Virus | |
IgG2a |
ELISA | |
JES3-19F1 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.