Learn More
IL-10 Rat anti-Mouse, FITC, Clone: JES5-16E3, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 562037
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The JES5-16E3 monoclonal antibody specifically binds to the mouse cytokine, Interleukin-10 (IL-10). IL-10 is also known as Cytokine Synthesis Inhibitory Factor (CSIF). It is produced by various activated cell types including CD4+ T cells, CD8+ T cells, T regulatory cells, NK T cells, B1 B cells, NK cells, macrophages, dendritic cells, mast cells, granulocytes and keratinocytes. IL-10 plays a pivotal role in regulating immune responses and protecting the host from damage caused by inflammatory and autoimmune responses. IL-10 has numerous biological activities including the inhibition of cytokine synthesis by activated T cells, NK cells, monocytes, and macrophages. In the presence of accessory cells, IL-10 inhibits mitogen- or anti-CD3 induced proliferation of T lymphocytes. IL-10 has also been shown to costimulate the development of thymocytes, B cell differentiation and the generation of cytotoxic T cells. The immunogen used to generate the JES5-16E3 hybridoma was recombinant mouse IL-10. JES5-16E3 is a neutralizing antibody.
Spezifikation
IL-10 | |
Monoclonal | |
0.5mg/mL | |
Interleukin-10; Il10; CSIF; Cytokine synthesis inhibitory factor | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b |
Flow Cytometry | |
JES5-16E3 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant mouse IL-10 | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.