Learn More
IL-17A Rat anti-Mouse, PE, Clone: TC11-18H10, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561020
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The TC11-18H10 monoclonal antibody specifically binds to recombinant and natural mouse IL-17A proteins. IL-17A, also known as CTLA-8, is a T cell-derived cytokine that promotes inflammatory responses. Mouse IL-17A is a proinflammatory cytokine that can induce the release of IL-6 by mouse stromal cells. It has been shown to support the growth of hemopoietic progenitors in vitro; it can also stimulate granulopoiesis in vivo. The TC11-18H10 antibody has been reported to neutralize IL-17A activity. Recent studies have shown that IL-17A is produced by a unique subset of Th17 cells that develop along a pathway distinct from the Th1- and Th2- cell differentiation pathways. The mouse IL-17A cDNA was isolated from a cDNA library generated from TCR αβ +CD4-CD8- thymocytes.
Spezifikation
IL-17A | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Mouse IL-17A Protein | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
TC11-18H10 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
For Research Use Only.