missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-27 Monoclonal Antibody (234205), PE, Invitrogen™
Rat Monoclonal Antibody
Marke: Thermo Scientific MA523651
Weitere Details : Netto-Gewicht : 0.00010kg
Description
This antibody recognizes the p28 subunit either as part of a p28/EBI-3 heterodimer or as a free subunit.
IL-27 Monoclonal antibody specifically detects IL-27 in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-27 | |
Monoclonal | |
PE | |
IL27 | |
cytokine receptor; EBI3; EBI-3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; Epstein-Barr virus induced gene 3 (similar to Interleukin-27 beta chain precursor); epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; Il27; IL-27; IL27 p28; IL-27 p28 subunit; IL27 subunit; IL-27 subunit alpha; IL-27 subunit beta; Il27a; IL-27A; IL-27-A; IL27-A; Il27b; IL-27B; IL27p28; IL-27p28; Il30; IL35 subunit; ILN; Interleukin; interleukin 27; interleukin 30; interleukin-27 subunit alpha; Interleukin-27 subunit beta; interleukin-30; MGC71873; M-IL-27 (p28); p28; RGD1561420 | |
Rat | |
Protein A/G | |
RUO | |
246779, 50498 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
234205 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
O35228, Q8K3I6 | |
Ebi3, IL27 | |
CHO cell-derived recombinant mouse IL-27 (aa 29-234 of p28, aa 19-228 of EBI-3) | |
100 Tests | |
Primary | |
Mouse | |
Antibody | |
IgG2b |