Learn More
IL-6 Recombinant Polyclonal Antibody (4HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710042
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
IL-6 Recombinant Polyclonal antibody specifically detects IL-6 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
IL-6 | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
Protein corresponding to amino acids 30-212 of human IL-6 | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
4HCLC | |
Unconjugated | |
P05231 | |
Rabbit | |
Protein A | |
RUO | |
3569 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |