Learn More
IL-6 Rat anti-Human, PE, Clone: MQ2-6A3, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 559331
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
The MQ2-6A3 antibody reacts with human interleukin-6 (IL-6). The immunogen used to generate the MQ2-6A3 hybridoma was recombinant human IL-6. This is a neutralizing antibody.
Intracellular Staining
Spezifikation
IL-6 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant human IL-6 | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Flow Cytometry | |
MQ2-6A3 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.