missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12 p40 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518741
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
IL-12 p40 Polyclonal antibody specifically detects IL-12 p40 in Mouse, Rat samples. It is validated for Western Blot.Spezifikation
IL-12 p40 | |
Polyclonal | |
Unconjugated | |
Il12b | |
CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; Il12; IL12 p40; IL-12 p40 subunit; IL-12 subunit p35; IL-12 subunit p40; IL12, subunit p40; IL-12.p40; IL12A; IL-12A; Il12b; IL-12B; Il12p40; IL-12p40; IL-12p40 subunit; ILN; IMD28; IMD29; Interleukin; Interleukin 12; interleukin 12 p40 subunit; interleukin 12 subunit p40; interleukin 12, p40; interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); Interleukin12; interleukin-12 beta chain; interleukin-12 beta chain precursor; interleukin-12 p40; interleukin-12 p40 chain; interleukin-12 p40 subunit; interleukin-12 subunit beta; interleukin-12 subunit p40; interleukin-12p40; interleukin-23 p40 subunit; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p40; RP23-388G23.1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16160, 64546 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P43432 | |
Il12b | |
Synthetic peptide sequence (EARQQNKYENYS) corresponding to the internal amino acids of l12b | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |