Learn More
Influenza A NP Monoclonal Antibody (I97D), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA17056
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-7056 detects the Influenza A virus. This antibody recognizes the nucleoprotein of numerous strains. MA1-7056 has been successfully used in immunofluorescence, immunohistochemistry and ELISA procedures. The MA1-7056 immunogen is Influenza A virus lysate.
Influenza A NP Monoclonal antibody specifically detects Influenza A NP in Virus samples. It is validated for ELISA, Immunohistochemistry.Spezifikation
Influenza A NP | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Immunohistochemistry | |
I97D | |
Unconjugated | |
Influenza nucleoprotein | |
Influenza A virus lysate. | |
1 mL | |
Primary | |
4° C | |
Liquid |