Learn More
Influenza A NP Monoclonal Antibody (A95I), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA17324
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The MA1-7324 antibody reacts with Influenza A virus NP in viral and infected tissue samples. MA1-7324 has been successfully used in Immunofluorescence and ELISA applications. The MA1-7324 antibody was raised against Influenza A Nucleoprotein.
Influenza A NP Monoclonal antibody specifically detects Influenza A NP in Virus samples. It is validated for ELISA, Immunocytochemistry.Spezifikation
Influenza A NP | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
A95I | |
Unconjugated | |
Influenza nucleoprotein | |
Influenza A Nucleoprotein. | |
100 μg | |
Primary | |
4° C | |
Liquid |