missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 HA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534920
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Influenza A H1N1 HA Polyclonal antibody specifically detects Influenza A H1N1 HA in Virus samples. It is validated for Western Blot.Spezifikation
Influenza A H1N1 HA | |
Polyclonal | |
Unconjugated | |
flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; Influenza H1N1; influenza hemagglutinin | |
Recombinant fragment corresponding to Internal region of Influenza A virus HA (A/duck/Alberta/35/1976(H1N1)) | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |