missing translation for 'onlineSavingsMsg'
Learn More

iNOS/NOS Type II Mouse anti-Mouse, FITC, Clone: 6, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610330

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15895399

  • 381.90 € / 1 Stück
Verfügbar ab: 02-01-2023
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits cellular signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In macrophages and other cell types, NOS (iNOS or macNOS) activity increases following exposure to cytokines (IFN-γ, TNF-α and IL-1) and microbial products (lipopolysaccharide (LPS)), iNOS is activated independently of Ca2+/calmodulin and its level of expression is tightly controlled by several transcription factors, including NFκB. Data indicates that TGF-β affects translation of iNOS mRNA and decreases iNOS protein stability. Normally undetectable in brain tissue, iNOS mRNA has been observed in CNS tissues of animals under experimental pathologic conditions. iNOS and nNOS share 51% amino acid homology with the greatest degree of divergence in the calmodulin binding domain.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation

Spezifikation

Spezifikation

iNOS/NOS Type II
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Mouse
IgG2a
Immunofluorescence
6
FITC
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.