Learn More
INSR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532878
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%); Mouse, Rat, Bovine, Dog, Elephant, Panda (94%); Bat, Horse, Pig (88%); Marmoset (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
INSR Polyclonal antibody specifically detects INSR in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
INSR | |
Polyclonal | |
Unconjugated | |
INSR | |
4932439J01Rik; CD 220; CD220; CD220 antigen; CD220 beta; CD221; D630014A15Rik; HHF 5; HHF5; IGF-I receptor; INSR; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR 1; IR beta; IR1; IR-1; IR-A; IR-B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P06213 | |
INSR | |
Synthetic 16 amino acid peptide from internal region of human Insulin Receptor. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |