Learn More
ITGA9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527771
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa. Predicted reactivity: Mouse (93%), Chicken (83%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane glycoproteins composed of an alpha chain and a beta chain that mediate cell-cell and cell-matrix adhesion. The protein encoded by this gene, when bound to the beta 1 chain, forms an integrin that is a receptor for VCAM1, cytotactin and osteopontin. Expression of this gene has been found to be upregulated in small cell lung cancers.Spezifikation
ITGA9 | |
Polyclonal | |
Unconjugated | |
Itga9 | |
(alpha)9; 2610002H11Rik; 6720458D17Rik; AI461869; ALPHA-RLC; D130073C02; D9Ertd428e; integrin alpha 9; integrin alpha-9; Integrin alpha-RLC; integrin subunit alpha 9; integrin, alpha 9; ITGA4L; Itga9; Itga9l; RLC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13797 | |
Itga9 | |
Recombinant fragment corresponding to a region within amino acids 1 and 255 of Human Integrin alpha 9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |