Learn More
IFNGR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527841
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, THP-1, HepG2, HepG2 membrane extract. Predicted reactivity: Rhesus Monkey (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IFNGR1 Polyclonal antibody specifically detects IFNGR1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IFNGR1 | |
Polyclonal | |
Unconjugated | |
IFNGR1 | |
antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; Ifgr; IFN-gamma receptor 1; IFN-gammaR; IFN-gamma-R1; IFN-gamma-R-alpha; Ifngr; IFNGR1; IMD27A; IMD27B; immune interferon receptor 1; INF-g receptor; interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain; Nktar | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3459 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P15260 | |
IFNGR1 | |
Recombinant protein fragment corresponding to a region within amino acids 206 and 436 of Human Interferon gamma Receptor 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |