missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCP Monoclonal Antibody (Hs-14), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110384
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
VCP Monoclonal antibody specifically detects VCP in Human, Mouse samples. It is validated for Flow Cytometry.Spezifikation
VCP | |
Monoclonal | |
FITC | |
VCP | |
15S Mg(2+)-ATPase p97 subunit; 3110001E05; AAA ATPase p97; ALS14; CDC48; cell division cycle gene CDC48; CMT2Y; epididymis luminal protein 220; epididymis secretory protein Li 70; HEL-220; HEL-S-70; homolog of yeast cdc48; IBMPFD; IBMPFD1; Inv protein; or HEL-S-70; p97; p97 atpase; p97/VCP; Protein CDC48; si:ch211-113n10.2; TER ATPase; TERA; transitional endoplasmic reticulum ATPase; valosin containing protein; valosin-containing protein; Vcp; wu:fd16d05; wu:fj63d11; yeast Cdc48p homolog | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
269523, 7415 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
Hs-14 | |
TBS with 0.2% BSA and 15mM sodium azide; pH 8.0 | |
P55072, Q01853 | |
VCP | |
Freshly ejaculated human sperms were washed in PBS and extracted in 3% acetic acid, 10% glycerol, 30 mM benzaminidine. The acid extract was dialyzed against 0.2% acetic acid and subsequently used for immunization. | |
50 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgM |