missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQCC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535902
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IQCC Polyclonal antibody specifically detects IQCC in Human samples. It is validated for Immunocytochemistry.Spezifikation
IQCC | |
Polyclonal | |
Unconjugated | |
IQCC | |
BC026978; IQ domain-containing protein C; IQ motif containing C; Iqcc; RP4-622L5.6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55721 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q4KMZ1 | |
IQCC | |
Recombinant fragment corresponding to a region within amino acids 49 and 321 of IQCC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |