missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA19058
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
IRF5 Polyclonal antibody specifically detects IRF5 in Human samples. It is validated for Western Blot.Spezifikation
IRF5 | |
Polyclonal | |
Unconjugated | |
IRF5 | |
AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3663 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q13568 | |
IRF5 | |
Synthetic peptide corresponding to the C terminus amino acids QGPWPMHPAGMQ. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |