missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITFG3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531403
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549. Predicted reactivity: Rhesus Monkey (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ITFG3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cell surface; extracellular exosome; integral component of membrane.Spezifikation
ITFG3 | |
Polyclonal | |
Unconjugated | |
FAM234A | |
C16orf9; Fam234a; family with sequence similarity 234 member A; family with sequence similarity 234, member A; gs19; integrin alpha FG-GAP repeat containing 3; ITFG3; Protein FAM234A; protein ITFG3; RGD1311484 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H0X4 | |
FAM234A | |
Recombinant fragment corresponding to a region within amino acids 36 and 325 of Human ITFG3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |