missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525433
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ITGA5 Polyclonal antibody specifically detects ITGA5 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
ITGA5 | |
Polyclonal | |
Unconjugated | |
Itga5 | |
2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
16402, 3678 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P08648, P11688 | |
Itga5 | |
KLH conjugated synthetic peptide between 796-822 amino acids from the C-terminal region of human ITGA5 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |