Learn More
IVD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522342
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, Jurkat, mouse brain, PC-12. Predicted reactivity: Mouse (90%), Rat (90%), Pig (91%), Chicken (85%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IVD Polyclonal antibody specifically detects IVD in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IVD | |
Polyclonal | |
Unconjugated | |
IVD | |
1300016K07Rik; 6720455E18Rik; ACAD2; AI463340; butyryl-CoA dehydrogenase; EGK_17362; isovaleryl CoA dehydrogenase; isovaleryl coenzyme A dehydrogenase; isovaleryl dehydrogenase; isovaleryl-CoA dehydrogenase; isovaleryl-CoA dehydrogenase, mitochondrial; IVD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24513, 3712, 56357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P12007, P26440, Q9JHI5 | |
IVD | |
Recombinant fragment corresponding to a region within amino acids 1 and 222 of IVD (Uniprot ID#P26440) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |