Learn More
IVD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544896
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: APKAQEIDRS NEFKNLREFW KQLGNLGVLG ITAPVQYGGS GLGYLEHVLV Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 86%
IVD Polyclonal antibody specifically detects IVD in Human samples. It is validated for Western Blot.Spezifikation
IVD | |
Polyclonal | |
Unconjugated | |
IVD | |
1300016K07Rik; 6720455E18Rik; ACAD2; AI463340; butyryl-CoA dehydrogenase; EGK_17362; isovaleryl CoA dehydrogenase; isovaleryl coenzyme A dehydrogenase; isovaleryl dehydrogenase; isovaleryl-CoA dehydrogenase; isovaleryl-CoA dehydrogenase, mitochondrial; IVD | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3712 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P26440 | |
IVD | |
Synthetic peptide directed towards the N-terminal of human IVD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |