Learn More
KARS Monoclonal Antibody (8G12C1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515426
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15426 targets KARS in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15426 immunogen is purified recombinant fragment of KARS(aa90-174) expressed in E. Coli.<br>.
KARS Monoclonal antibody specifically detects KARS in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
KARS | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q15046 | |
KARS1 | |
Purified recombinant fragment of KARS(aa90-174) expressed in E. Coli. | |
RUO | |
3735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
8G12C1 | |
Unconjugated | |
KARS1 | |
AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |