missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCC1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518815
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
KCC1 Polyclonal antibody specifically detects KCC1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
KCC1 | |
Polyclonal | |
Unconjugated | |
SLC12A4 | |
AW546649; CTC-479C5.17; Electroneutral potassium-chloride cotransporter 1; erythroid K-Cl cotransporter 1; FLJ40489; Furosemide-sensitive K-Cl cotransporter; hKCC1; Kcc1; K-Cl cotransporter; K-Cl cotransporter 1; K-Cl cotransporter 4; K-Cl cotransporter KCC1; K-Cl Co-transporter-1; mKCC1; potassium chloride cotransporter SLC12A4; potassium/chloride cotransporter 1; potassium-chloride cotransporter-1; RBCKCC1; RBKCC1; rKCC1; Slc12a4; solute carrier family 12 (potassium/chloride transporter), member 4; solute carrier family 12 (potassium/chloride transporters), member 4; solute carrier family 12 member 4; solute carrier family 12, member 4 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6560 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UP95 | |
SLC12A4 | |
Synthetic peptide sequence (DPSHAPDNFRE) corresponding to the internal amino acids of SLC12A4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |