Learn More
KCNE3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533864
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Hamster (94%) Mouse, Bat (89%) Marmoset, Rat, Elephant, Panda, Dog, Bovine, Pig (83%).
KCNE3 Polyclonal antibody specifically detects KCNE3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
KCNE3 | |
Polyclonal | |
Unconjugated | |
Kcne3 | |
2210017H05Rik; cardiac voltage-gated potassium channel accessory subunit; HOKPP; HYPP; KCNE3; Minimum potassium ion channel-related peptide 2; minK-related peptide 2; MiRP2; potassium channel subunit beta MiRP2; potassium channel, voltage gated subfamily E regulatory beta subunit 3; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 3; potassium voltage-gated channel subfamily E member 3; potassium voltage-gated channel subfamily E regulatory subunit 3; potassium voltage-gated channel, Isk-related family, member 3; potassium voltage-gated channel, Isk-related subfamily, gene 3; potassium voltage-gated channel, Isk-related subfamily, member 3; voltage-gated K+ channel subunit MIRP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10008 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9Y6H6 | |
Kcne3 | |
Synthetic 18 amino acid peptide from internal region of human KCNE3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |