Learn More
Kir6.1 (KCNJ8) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541028
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: EKPSILIQTL QKSELSHQNS LRKRNSMRRN NSMRRNNSIR RNNSSLMVPK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
Kir6.1 (KCNJ8) Polyclonal antibody specifically detects Kir6.1 (KCNJ8) in Human samples. It is validated for Western Blot.Spezifikation
Kir6.1 (KCNJ8) | |
Polyclonal | |
Unconjugated | |
KCNJ8 | |
AI448900; ATP-sensitive inward rectifier potassium channel 8; ATP-sensitive inward rectifier potassium channel Kir6.1; gnite; Inward rectifier K(+) channel Kir6.1; inward rectifier potassium channel 6.1; inward rectifier potassium channel Kir6.1; inwardly rectifier K(+) channel Kir6.1; Inwardly rectifying potassium channel gene subfamily J-8 (ATP sensitive); Inwardly rectifying potassium channel gene, subfamily J-8 (ATP sensitive); inwardly rectifying potassium channel Kir6.1; ion channel; Kcnj8; KCNJ8 protein; Kir6.1; Kir6.1 like; potassium channel, inwardly rectifying subfamily J member 8; potassium channel, inwardly rectifying subfamily J, member 8; potassium inwardly rectifying channel subfamily J member 8; potassium inwardly-rectifying channel J8; potassium inwardly-rectifying channel, subfamily J, member 8; potassium voltage-gated channel subfamily J member 8; si:dkey-183c2.2; slmbr; sltr; UKATP1; uKATP-1; zkir6.1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3764 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15842 | |
KCNJ8 | |
Synthetic peptide directed towards the middle region of human KCNJ8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |