Learn More
KCNK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541010
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: EDQVHIIEHD QLSFSSITDQ AAGMKEDQKQ NEPFVATQSS ACVDGPANH Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 100%; Rat: 93%
KCNK1 Polyclonal antibody specifically detects KCNK1 in Human samples. It is validated for Western Blot.Spezifikation
KCNK1 | |
Polyclonal | |
Unconjugated | |
Kcnk1 | |
AI788889; double-pore potassium channel; DPK; HOHO; HOHO1; Inward rectifying potassium channel protein TWIK-1; K2P1; K2p1.1; Kcnk1; kcnk1 {ECO:0000312; KCNO1; Potassium channel K2P1; potassium channel KCNO1; Potassium channel subfamily K member 1; potassium channel TWIK-1; potassium channel, subfamily K, member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; potassium two pore domain channel subfamily K member 1; putative potassium channel TWIK; rabKCNK1; RGD:621447}; rTWIK; tandem of P domains in a weak inward rectifying K+ channel 1; Twik; TWIK1; TWIK-1; two-pore potassium channel; Unknown (protein for MGC:142788) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3775 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00180 | |
Kcnk1 | |
Synthetic peptide directed towards the middle region of human KCNK1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |