Learn More
KCTD19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532114
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: U87-MG, SK-N-SH. Predicted reactivity: Mouse (96%), Rat (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KCTD19 Polyclonal antibody specifically detects KCTD19 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
KCTD19 | |
Polyclonal | |
Unconjugated | |
KCTD19 | |
4922504H04Rik; BTB/POZ domain-containing protein KCTD19; KCTD19; potassium channel tetramerisation domain containing 19; potassium channel tetramerization domain containing 19; RGD1561326; testicular tissue protein Li 101 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146212, 279499 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q17RG1, Q562E2 | |
KCTD19 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 162 of Human KCTD19 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |