Learn More
JMJD1A Monoclonal Antibody (2E1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515724
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15724 targets KDM3A in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15724 immunogen is purified recombinant fragment of human KDM3A expressed in E. Coli. MA5-15724 detects KDM3A which has a predicted molecular weight of approximately 147kDa.
JMJD1A Monoclonal antibody specifically detects JMJD1A in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
JMJD1A | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q9Y4C1 | |
KDM3A | |
Purified recombinant fragment of human KDM3A expressed in E. Coli. | |
RUO | |
55818 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
2E1 | |
Unconjugated | |
KDM3A | |
1700105C21Rik; C230043E16Rik; DKFZp686A24246; DKFZp686P07111; JHDM2a; JHMD2A; JmjC domain-containing histone demethylation protein 2A; Jmjd1; Jmjd1a; jumonji C domain-containing histone demethylase 2A; jumonji domain containing 1A; jumonji domain-containing protein 1A; KDM2A; Kdm3a; Kiaa0742; lysine (K)-specific demethylase 3A; lysine demethylase 3A; lysine-specific demethylase 3A; probable zinc finger protein; testis specific gene A; testis-specific gene A protein; testis-specific protein A; TGSA; Tsga; zinc finger protein TSGA | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |